Learn More
Invitrogen™ ABCA4 Monoclonal Antibody (3F4)

Description
Excitation: 405 nm; Emission: 645 nm; Laser: Blue Laser.
Specifications
Specifications
| Antigen | ABCA4 |
| Applications | Immunohistochemistry, Western Blot |
| Classification | Monoclonal |
| Clone | 3F4 |
| Concentration | ∼1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | 10mM HEPES with 100μg/mL BSA, 50% glycerol, 0.15M NaCl and no preservative; pH 7.5 |
| Gene | ABCA4 |
| Gene Accession No. | O35600, P78363 |
| Gene Alias | ABC transporter; ABC10; abca4; abca4.L; Abcr; ARMD2; ATP binding cassette subfamily A member 4; ATP binding cassette subfamily A member 4 L homeolog; ATP binding cassette transporter; ATP-binding cassette 10; ATP-binding cassette sub-family A member 4; ATP-binding cassette transporter, retinal-specific; ATP-binding cassette, subfamily A (ABC1), member 4; ATP-binding cassette, sub-family A (ABC1), member 4; ATP-binding cassette, sub-family A, member 4; ATP-binding transporter, retina-specific; AW050280; CORD3; D430003I15Rik; FFM; photoreceptor rim protein; retinal ABCA4 transporter; retinal-specific ATP-binding cassette transporter; Retinal-specific phospholipid-transporting ATPase ABCA4; retina-specific ABC transporter; RIM ABC transporter; RIM protein; RMP; RP19; Stargardt disease protein; STGD; STGD1; XELAEV_180233141mg |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.