Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ABCA4 Mouse anti-Bovine, Mouse, Clone: 3F4, Abnova™

Catalog No. 89035376 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-035-376 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-035-376 Supplier Abnova Corporation Supplier No. MAB13969

Mouse Monoclonal Antibody

Mouse monoclonal antibody raised against native bovine ABCA4.

  • High specificity
  • High reactivity

Specifications

Antigen ABCA4
Applications Immunohistochemistry (PFA fixed), Western Blot
Classification Monoclonal
Clone 3F4
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against native bovine ABCA4.
Dilution Immunohistochemistry (1:100) Western Blot (1:1000) The optimal working dilution should be determined by the end user.
Formulation In 0.01M HEPES, 0.15 M NaCl, pH 7.5 (100 ug/mL BSA, 50% glycerol).
Gene Symbols ABCA4
Host Species Mouse
Immunogen Native purified bovine ABCA4.
Purification Method Protein G purification
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 281584
Target Species Bovine, Mouse
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.