Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ABCA4 Mouse anti-Human, Bovine, Unconjugated, Clone: 5B4, MilliporeSigma™

Catalog No. MABN244025 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABN2440-25 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABN2440-25 Supplier MilliporeSigma Supplier No. MABN244025UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-ABCA4, clone 5B4, Cat. No. MABN2440, is a highly specific mouse monoclonal antibody that targets Retinal-specific ATP-binding cassette transporter and has been tested for use in Immunocytochemistry, Immunoprecipitation, and Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ABCA4
Applications Immunocytochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 5B4
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Formulation Purified mouse monoclonal antibody IgG2b in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.
Gene Alias Retinal-specific ATP-binding cassette transporter;ATP-binding cassette sub-family A member 4;RIM ABC transporter;RIM protein;RmP;Stargardt disease protein
Gene Symbols ABCA4;ABCR
Host Species Mouse
Immunogen GST-tagged recombinant fragment corresponding to 249 amino acids from the N-terminal half of human Retinal-specific ATP-binding cassette transporter.
Molecular Weight of Antigen 250 kDa observed; 255.94 kDa calculated. Uncharacterized bands may be observed in some lysate(s).
Purification Method Protein G purified
Quantity 25 μg
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Test Specificity Clone 5B4 detects Retinal-specific ATP-binding cassette transporter protein in human and bovine. It targets an epitope within 249 amino acids from the N-terminal half.
Target Species Human, Bovine
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Product Type Antibodies
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.