Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ ABCA4, Mouse, Unlabeled, Clone: TMR1,

Catalog No. MABN1775MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
200 μL
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
MABN1775MI 200 μL Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABN1775MI Supplier MilliporeSigma™ Supplier No. MABN1775
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Retinal-specific ATP-binding cassette transporter (UniProt F1MWM0/P78363; also known as ATP-binding cassette sub-family A member 4, ABC transporter A4, RIM ABC transporter, RIM protein, RmP, Stargardt disease protein) is encoded by the ABCA4 (also known as ABCR) gene (Gene ID 281584/24) in bovine/human species. ABC transporter A4, a ∽250kDa single-chain, is a multi-pass membrane protein of the superfamily of ATP-binding cassette (ABC) transporters that is localized to the disk margins of vertebrate photoreceptor outer segments. It is implicated in the clearance of all-trans retinal derivatives from rod and cone photoreceptor cells following photoexcitation as part of the visual cycle. ABC transporter A4 is composed of two symmetrically organized halves, each comprising six membrane-spanning helices, a large glycosylated exocytoplasmic domain located inside the disk, and a cytoplasmic domain with an ATP-binding cassette. Mutations in ABCA4 are known to cause Stargardt macular degeneration, which is an autosomal recessive form of juvenile macular degeneration associated with a severe loss in vision. Ref.: Tsybovsky, Y et al. (2010). Adv. Exp. Med. Biol. 703, 105-125.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ABCA4
Applications Immunofluorescence, Western Blot
Classification Monoclonal
Clone TMR1
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Formulation Purified mouse IgG1 in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% Sodium Azide.
Gene ABCA4, ABCR
Gene Accession No. F1MWM0
Host Species Mouse
Immunogen ABCA4 isolated from bovine retinas (Zhang, N., et al. (2015). Hum. Mol. Genet. 24(11):3220-3237).
Purification Method Protein G Purified
Quantity 200 μL
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) NP_776646
Test Specificity Clone TMR1 immunostained retina cryosections from wild-type mice, but not retina from mice with homozygous L541P;A1038V (PV) missense mutation (Courtesy of Grace Jin, Case Western University, U.S.A.).
Target Species Human
Content And Storage Stable for 1 year at 2°-8°C from date of receipt.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.