Learn More
Invitrogen™ ABCA4 Polyclonal Antibody

Description
Positive Samples: U-251MG, BT-474, 293T, Mouse heart, Mouse lung, Rat eye; Cellular Location: Membrane, Multi-pass membrane protein Immunogen sequence: QELLMNAPES QHLGRIWTEL HILSQFMDTL RTHPERIAGR GIRIRDILKD EETLTLFLIK NIGLSDSVVY LLINSQVRPE QFAHGVPDLA LKDIACSEAL LERFIIFSQR RGAKTVRYAL CSLSQGTLQW IEDTLYANVD FFKLFRVLPT LLDSRSQGIN LRSWGGILSD MSPRIQEFIH RPSMQDLLWV T.
Specifications
Specifications
| Antigen | ABCA4 |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Concentration | 1.57 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3 |
| Gene | ABCA4 |
| Gene Accession No. | O35600, P78363 |
| Gene Alias | ABC transporter; ABC10; abca4; abca4.L; Abcr; ARMD2; ATP binding cassette subfamily A member 4; ATP binding cassette subfamily A member 4 L homeolog; ATP binding cassette transporter; ATP-binding cassette 10; ATP-binding cassette sub-family A member 4; ATP-binding cassette transporter, retinal-specific; ATP-binding cassette, subfamily A (ABC1), member 4; ATP-binding cassette, sub-family A (ABC1), member 4; ATP-binding cassette, sub-family A, member 4; ATP-binding transporter, retina-specific; AW050280; CORD3; D430003I15Rik; FFM; photoreceptor rim protein; retinal ABCA4 transporter; retinal-specific ATP-binding cassette transporter; Retinal-specific phospholipid-transporting ATPase ABCA4; retina-specific ABC transporter; RIM ABC transporter; RIM protein; RMP; RP19; Stargardt disease protein; STGD; STGD1; XELAEV_180233141mg |
| Gene Symbols | ABCA4 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.