Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ABCA4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5142378
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5142378 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5142378 Supplier Invitrogen™ Supplier No. PA5142378
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:128,000.

ABCA4 (ATP-Binding Cassette, Subfamily A, Member 4), also known as ABCR, is a protein which in humans is encoded by the ABCA4 gene. ABCA4 is a member of the ATP-binding cassette transporter gene sub-family A (ABC1) found exclusively in multicellular eukaryotes. Using a whole genome radiation hybrid panel, this gene is mapped to 1p21-p13. And this gene is expressed exclusively in retina photoreceptor cells, indicating the gene product mediates transport of an essental molecule across the photoreceptor cell membrane. Additionally, it is showed by immunofluorescence microscopy and Western blot analysis that ABCR is present in foveal and peripheral cone, as well as rod, photoreceptors. The results suggested that the loss in central vision experienced by patients with Stargardt macular dystrophy arises directly from ABCR-mediated foveal cone degeneration.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ABCA4
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene ABCA4
Gene Accession No. O35600, P78363
Gene Alias ABC transporter; ABC10; abca4; abca4.L; Abcr; ARMD2; ATP binding cassette subfamily A member 4; ATP binding cassette subfamily A member 4 L homeolog; ATP binding cassette transporter; ATP-binding cassette 10; ATP-binding cassette sub-family A member 4; ATP-binding cassette transporter, retinal-specific; ATP-binding cassette, subfamily A (ABC1), member 4; ATP-binding cassette, sub-family A (ABC1), member 4; ATP-binding cassette, sub-family A, member 4; ATP-binding transporter, retina-specific; AW050280; CORD3; D430003I15Rik; FFM; photoreceptor rim protein; retinal ABCA4 transporter; retinal-specific ATP-binding cassette transporter; Retinal-specific phospholipid-transporting ATPase ABCA4; retina-specific ABC transporter; RIM ABC transporter; RIM protein; RMP; RP19; Stargardt disease protein; STGD; STGD1; XELAEV_180233141mg
Gene Symbols ABCA4
Host Species Goat
Immunogen Peptide with sequence C-KQQTESHDLPLHPR.
Purification Method Ammonium sulfate precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11304, 24
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.