Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ABCA5 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA537131
Description
This antibody detects endogenous protein at a molecular weight of 186 kDa. Purity is >95% by SDS-PAGE.
ABCA5 (ATP-binding cassette, sub-family A (ABC1), member 5) is a 1,642 amino acid protein belonging to the ABC transporter superfamily and the ABCA family. The ABC1 subfamily is the only major ABC subfamily exclusive to multicellular eukaryotes. Ubiquitously expressed, with high expression in testis, skeletal muscle, kidney, liver and placenta, ABCA5 is a multi-pass membrane protein that contains two ABC transporter domains and exists as three alternatively spliced isoforms. ABCA5 exhibits membrane subcellular localization and may play a role in processing autolysosomes. Spanning 80 kb, ABCA5 contains 39 exons, with exon 2 containing the putative translation start site. ABCA5 is the first of five ABC1 family members that maps to human chromosome 17q24. ABCA5 is linked to lysosomal diseases and may play a role in tumor development and cardiomyocyte and follicular cell activities. ABCA5 might also be a specific urine marker for diagnosis of patients with high-grade prostatic intraepithelial neoplasia (HGPIN).
Specifications
| ABCA5 | |
| Polyclonal | |
| Unconjugated | |
| ABCA5 | |
| ABC13; Abca5; ATP binding cassette subfamily A member 5; ATP-binding cassette A5; ATP-binding cassette sub-family A member 5; ATP-binding cassette transporter sub-family A member 5; ATP-binding cassette, subfamily A (ABC1), member 5; ATP-binding cassette, sub-family A (ABC1), member 5; ATP-binding cassette, sub-family A , member 5; B930033A02Rik; DKFZp451F117; DKFZp779N2435; EST90625; FLJ16381; Kiaa1888; mKIAA1888 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 217265, 23461, 286970 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8CF82, Q8K448, Q8WWZ7 | |
| ABCA5 | |
| A synthetic peptide corresponding to residues in human ABCA5. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction