Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ABCA7 Antibody (7A1-144), Biotin, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals NB400163B
Description
ABCA7 Monoclonal specifically detects ABCA7 in Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
ABCA7 | |
Monoclonal | |
Biotin | |
ABCA-SSN, ABCXATP-binding cassette sub-family A member 7, ATP-binding cassette, sub-family A (ABC1), member 7, Autoantigen SS-N, EC 2.2.1.1, EC 3.6.3, EC 3.6.3.41, FLJ40025, Macrophage ABC transporter | |
Rat | |
240 kDa | |
0.1 mL | |
Astrocyte Markers, Cellular Markers, Cholesterol Metabolism, Glia Markers, Golgi Apparatus Markers, Lipid and Metabolism, Neuroscience, Plasma Membrane Markers, Signal Transduction | |
10347 | |
Store at 4C in the dark. | |
IgG2a κ |
Western Blot, Immunohistochemistry (Paraffin) | |
7A1-144 | |
Western Blot, Immunohistochemistry-Paraffin | |
ABCA7 | |
Mouse ABCA7 stably-transfected HeLa cells. [UniProt# Q91V24] | |
Protein G purified | |
RUO | |
Primary | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction