Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ABCA7 Antibody (7A1-144) - Unpurified, Novus Biologicals™

Rat Monoclonal Antibody
$206.50 - $426.00
Specifications
Antigen | ABCA7 |
---|---|
Clone | 7A1-144 |
Dilution | Western Blot 1:2000-1:8000, Flow Cytometry 1:10000-1:60000, Immunohistochemistry-Paraffin 1:100-1:250 |
Applications | Western Blot, Flow Cytometry, Immunohistochemistry (Paraffin) |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB400166SS
![]() |
Novus Biologicals
NB400166SS |
0.025 mL |
Each for $206.50
|
|
|||||
NB400166
![]() |
Novus Biologicals
NB400166 |
0.1 mL |
Each for $426.00
|
|
|||||
Description
ABCA7 Monoclonal specifically detects ABCA7 in Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
ABCA7 | |
Western Blot 1:2000-1:8000, Flow Cytometry 1:10000-1:60000, Immunohistochemistry-Paraffin 1:100-1:250 | |
Monoclonal | |
Ascites | |
RUO | |
Mouse | |
ABCA-SSN, ABCXATP-binding cassette sub-family A member 7, ATP-binding cassette, sub-family A (ABC1), member 7, Autoantigen SS-N, EC 2.2.1.1, EC 3.6.3, EC 3.6.3.41, FLJ40025, Macrophage ABC transporter | |
ABCA7 | |
IgG2a κ | |
Unpurified | |
240 kDa |
7A1-144 | |
Western Blot, Flow Cytometry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rat | |
ABC Transporters, Astrocyte Markers, Cellular Markers, Cholesterol Metabolism, Glia Markers, Golgi Apparatus Markers, Lipid and Metabolism, Neuroscience, Plasma Membrane Markers, Signal Transduction | |
Ascites with 0.1% Sodium Azide | |
10347 | |
Mouse ABCA7 stably-transfected HeLa cells. [UniProt# Q91V24] | |
Primary | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title