Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ABCA8 Polyclonal Antibody

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA550902

Catalog No. PA550902


Only null left
Add to Cart

Description

Description

ABCA8 Polyclonal Antibody detects endogenous levels of ABCA8 protein.

ATP-binding cassette (ABC) transporters are an evolutionarily conserved family of proteins that catalyze the transport of molecules across extracellular and intracellular membranes by harnessing the energy of ATP hydrolysis. ABCA8 (ATP-binding cassette, sub-family A (ABC1), member 8) is a 1, 581 amino acid multi-pass membrane protein that contains two ABC transporter domains and belongs to the ABC transporter family. Expressed in a variety of tissues with particularly high expression in liver, heart and skeletal muscle, ABCA8 functions as an ATP-dependent lipophilic drug transporter that exists as multiple alternatively spliced isoforms.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

ABCA8
Polyclonal
Unconjugated
ABCA8
Abca8; Abca8a; Abca8b; ATP binding cassette subfamily A member 8; ATP-binding cassette sub-family A member 8; ATP-binding cassette sub-family A member 8-A; ATP-binding cassette sub-family A member 8-B; ATP-binding cassette, subfamily A (ABC1), member 8; ATP-binding cassette, sub-family A (ABC1), member 8; ATP-binding cassette, subfamily A (ABC1), member 8a; ATP-binding cassette, sub-family A (ABC1), member 8a; ATP-binding cassette, sub-family A (ABC1), member 8b; KIAA0822
Rabbit
Antigen Affinity Chromatography
RUO
10351, 217258, 27404
-20°C, Avoid Freeze/Thaw Cycles
Liquid
ELISA, Western Blot
1 mg/mL
PBS with 0.5% BSA, 50% glycerol and 0.02% sodium azide; pH 7.4
O94911, Q8K440, Q8K442
ABCA8, Abca8a, Abca8b
Synthesized peptide derived from the Internal region of human ABCA8.
100 μL
Primary
Human, Mouse
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.