Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ABCA8 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA560866
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA560866 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA560866 Supplier Invitrogen™ Supplier No. PA560866
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: KEVLGLPDEE SIKEFTANYP EEIVRVTFTN TYSYHLKFLL GHGMPAKKEH KDHTAHCYET NEDVYCEVSV FWKEGFVA Highest antigen sequence identity to the following orthologs: Mouse - 55%, Rat - 55%.

ATP-binding cassette (ABC) transporters are an evolutionarily conserved family of proteins that catalyze the transport of molecules across extracellular and intracellular membranes by harnessing the energy of ATP hydrolysis. ABCA8 (ATP-binding cassette, sub-family A (ABC1), member 8) is a 1, 581 amino acid multi-pass membrane protein that contains two ABC transporter domains and belongs to the ABC transporter family. Expressed in a variety of tissues with particularly high expression in liver, heart and skeletal muscle, ABCA8 functions as an ATP-dependent lipophilic drug transporter that exists as multiple alternatively spliced isoforms.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ABCA8
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ABCA8
Gene Accession No. O94911
Gene Alias Abca8; Abca8a; Abca8b; ATP binding cassette subfamily A member 8; ATP-binding cassette sub-family A member 8; ATP-binding cassette sub-family A member 8-A; ATP-binding cassette sub-family A member 8-B; ATP-binding cassette, subfamily A (ABC1), member 8; ATP-binding cassette, sub-family A (ABC1), member 8; ATP-binding cassette, subfamily A (ABC1), member 8a; ATP-binding cassette, sub-family A (ABC1), member 8a; ATP-binding cassette, sub-family A (ABC1), member 8b; KIAA0822
Gene Symbols ABCA8
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ABCA8. Recombinant protein control fragment (Product #RP-93289).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10351
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.