Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ABCA9 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA518953
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA518953 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA518953 Supplier Invitrogen™ Supplier No. PA518953
Add to Cart
Add to Cart

Goat Polyclonal Antibody

This antibody is tested in Peptide ELISA: antibody detection limit dilution 128,000.

ABCA9 (ATP-binding cassette, sub-family A (ABC1), member 9) is a 1,624 amino acid protein belonging to the ABC transporter superfamily and the ABCA family. The ABC1 subfamily is the only major ABC subfamily exclusive to multicellular eukaryotes. Ubiquitously expressed, with high expression in heart, brain and fetal tissues, ABCA9 is a multi-pass membrane protein that contains two ABC transporter domains and exists as five alternatively spliced isoforms. ABCA9 exhibits membrane subcellular localization and may play a role in monocyte differentiation and lipid homeostasis. ABCA9 is up-regulated during monocyte differentiation into macrophages and down-regulated by cholesterol loading of macrophages. Spanning 85 kb, ABCA9 contains 39 exons, with a non-coding first exon.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ABCA9
Applications Flow Cytometry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene ABCA9
Gene Accession No. Q8IUA7
Gene Alias ABCA9; ATP binding cassette subfamily A member 9; ATP-binding cassette A9; ATP-binding cassette sub-family A member 9; ATP-binding cassette transporter sub-family A member 9; ATP-binding cassette, subfamily A (ABC1), member 9; ATP-binding cassette, sub-family A (ABC1), member 9; D630040K07Rik; DKFZp686F2450; EST640918; MGC75415
Gene Symbols ABCA9
Host Species Goat
Immunogen Synthetic peptide sequence (QRVVQELEMENIQD) corresponding to the internal amino acids of ABCA9 (aa 599-613).
Purification Method Ammonium Sulfate Precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10350
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.