Learn More
ABCB1 Mouse anti-Human, Clone: OTI5E3, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF801010

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
The membrane-associated protein encoded by this gene is a member of the superfamily of ATP-binding cassette (ABC) transporters. ABC proteins transport various molecules across extra- and intra-cellular membranes. ABC genes are divided into seven distinct subfamilies (ABC1, MDR/TAP, MRP, ALD, OABP, GCN20, White). This protein is a member of the MDR/TAP subfamily. Members of the MDR/TAP subfamily are involved in multidrug resistance. The protein encoded by this gene is an ATP-dependent drug efflux pump for xenobiotic compounds with broad substrate specificity. It is responsible for decreased drug accumulation in multidrug-resistant cells and often mediates the development of resistance to anticancer drugs. This protein also functions as a transporter in the blood-brain barrier.Specifications
ABCB1 | |
Monoclonal | |
Unconjugated | |
ABCB1 | |
ABC20, CD243, CLCS, GP170, MDR1, P-GP, PGY1 | |
Mouse | |
Affinity Chromatography | |
RUO | |
5243 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Western Blot | |
OTI5E3 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
P08183 | |
ABCB1 | |
Human recombit protein fragment corresponding to amino acids 995-1280 of human ABCB1 produced in SF9 cell. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.