Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ABCB10 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA575389
Description
The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).
The membrane-associated protein encoded by this gene is a member of the superfamily of ATP-binding cassette (ABC) transporters. ABC proteins transport various molecules across extra- and intra-cellular membranes. ABC genes are divided into seven distinct subfamilies (ABC1, MDR/TAP, MRP, ALD, OABP, GCN20, White). This protein is a member of the MDR/TAP subfamily. Members of the MDR/TAP subfamily are involved in multidrug resistance. The function of this mitochondrial protein is unknown.
Specifications
ABCB10 | |
Polyclonal | |
Unconjugated | |
ABCB10 | |
ABC transporter 10 protein; ABCB10; Abcb12; Abc-me; ABC-me protein; ABC-mitochondrial erythroid protein; ATP binding cassette subfamily B member 10; ATP-binding cassette sub-family B member 10, mitochondrial; ATP-binding cassette transporter 10; ATP-binding cassette, subfamily B (MDR/TAP), member 10; ATP-binding cassette, sub-family B (MDR/TAP), member 10; EST20237; M-ABC2; mitochondrial ATP-binding cassette 2; MTABC2; si:dkey-162b3.3 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
23456, 361439, 56199 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9JI39, Q9NRK6 | |
ABCB10 | |
Synthetic peptide corresponding to amino acids 445-494 of Human ABCB10. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction