Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ABCB10 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5114799
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5114799 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5114799 Supplier Invitrogen™ Supplier No. PA5114799
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total ABCB10.

The membrane-associated protein encoded by this gene is a member of the superfamily of ATP-binding cassette (ABC) transporters. ABC proteins transport various molecules across extra- and intra-cellular membranes. ABC genes are divided into seven distinct subfamilies (ABC1, MDR/TAP, MRP, ALD, OABP, GCN20, White). This protein is a member of the MDR/TAP subfamily. Members of the MDR/TAP subfamily are involved in multidrug resistance. The function of this mitochondrial protein is unknown.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ABCB10
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide
Gene ABCB10
Gene Accession No. Q9NRK6
Gene Alias ABC transporter 10 protein; ABCB10; Abcb12; Abc-me; ABC-me protein; ABC-mitochondrial erythroid protein; ATP binding cassette subfamily B member 10; ATP-binding cassette sub-family B member 10, mitochondrial; ATP-binding cassette transporter 10; ATP-binding cassette, subfamily B (MDR/TAP), member 10; ATP-binding cassette, sub-family B (MDR/TAP), member 10; EST20237; M-ABC2; mitochondrial ATP-binding cassette 2; MTABC2; si:dkey-162b3.3
Gene Symbols ABCB10
Host Species Rabbit
Immunogen A synthesized peptide derived from human ABCB10(Accession Q9NRK6), corresponding to amino acid residues G446-N496.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23456, 361439
Target Species Human, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.