Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ABCB11 Antibody (IHC518), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP2759800.1ML
Description
ABCB11 Monoclonal specifically detects ABCB11 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin, Knockdown Validated.Specifications
ABCB11 | |
Monoclonal | |
Unconjugated | |
Tris Buffer, pH 7.3 - 7.7, with 1% BSA with 0.1% sodium azide | |
ABCB11 | |
Recombinant Human BSEP | |
Primary | |
Human | |
IgG1 |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
IHC518 | |
Immunohistochemistry 1:100 - 1:200, Immunohistochemistry-Paraffin 1:100 - 1:200 | |
ABC member 16, MDR/TAP subfamily, ABC16, ATP-binding cassette sub-family B member 11, ATP-binding cassette, sub-family B (MDR/TAP), member 11, bile salt export pump, BSEP, BSEPPFIC2, EC 3.6.3, EC 3.6.3.44, PFIC-2, PGY4, progressive familial intrahepatic cholestasis 2, sister p-glycoprotein, SPGPBRIC2 | |
Mouse | |
0.1 mL | |
8647 | |
Store at 4°C in the dark. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction