Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ABCB11 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5103659
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5103659 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5103659 Supplier Invitrogen™ Supplier No. PA5103659
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total ABCB11.

ABCB11 is involved in the ATP-dependent secretion of bile salts into the canaliculus of hepatocytes. It is expressed predominatly, if not exclusively, in the liver, where it is further localized to the canilicular microvilli and to subcanilicular vesicles fo the hepatocytes. Structurally, ABCB11 is a multifunctional polypeptide with two homologus halves, each containing a hydrophobic membrane-anchoring domain and an ATP binding cassette (ABC) domain. Defects in ABCB11 are the cause of progressive familial intrahepatic cholestasis 2 (PFIC2). PFIC2 is an inherited liver disease of childhood which is characterized by cholestasis and normal serum gamma-glutamyltransferase activity. Defects in ABCB11 are also found in cases of chronic intrahepatic cholestasis without obvious familial history of chronic liver disease.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ABCB11
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene ABCB11
Gene Accession No. O70127, O95342, Q9QY30
Gene Alias ABC member 16, MDR/TAP subfamily; ABC16; Abcb11; ATP binding cassette subfamily B member 11; ATP-binding cassette sub-family B member 11; ATP-binding cassette, subfamily B (MDR/TAP), member 11; ATP-binding cassette, sub-family B (MDR/TAP), member 11; ATP-binding cassette, sub-family B, member 11; bile salt export pump; BRIC2; Bsep; Lith1; PFIC2; PFIC-2; PGY4; progressive familial intrahepatic cholestasis 2; sister of P-glycoprotein; sister p-glycoprotein; Spgp
Gene Symbols ABCB11
Host Species Rabbit
Immunogen A synthesized peptide derived from human ABCB11(Accession O95342), corresponding to amino acid residues N665-V715.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27413, 83569, 8647
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.