Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ BSEP Polyclonal Antibody

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA513105

Catalog No. PIPA513105


Only null left
Add to Cart

Description

Description

BSEP Polyclonal Antibody for Western Blot, IHC (P)

ABCB11 is involved in the ATP-dependent secretion of bile salts into the canaliculus of hepatocytes. It is expressed predominatly, if not exclusively, in the liver, where it is further localized to the canilicular microvilli and to subcanilicular vesicles fo the hepatocytes. Structurally, ABCB11 is a multifunctional polypeptide with two homologus halves, each containing a hydrophobic membrane-anchoring domain and an ATP binding cassette (ABC) domain. Defects in ABCB11 are the cause of progressive familial intrahepatic cholestasis 2 (PFIC2). PFIC2 is an inherited liver disease of childhood which is characterized by cholestasis and normal serum gamma-glutamyltransferase activity. Defects in ABCB11 are also found in cases of chronic intrahepatic cholestasis without obvious familial history of chronic liver disease.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

BSEP
Polyclonal
Unconjugated
ABCB11
ABC member 16, MDR/TAP subfamily; ABC16; Abcb11; ATP binding cassette subfamily B member 11; ATP-binding cassette sub-family B member 11; ATP-binding cassette, subfamily B (MDR/TAP), member 11; ATP-binding cassette, sub-family B (MDR/TAP), member 11; ATP-binding cassette, sub-family B, member 11; bile salt export pump; BRIC2; Bsep; Lith1; PFIC2; PFIC-2; PGY4; progressive familial intrahepatic cholestasis 2; sister of P-glycoprotein; sister p-glycoprotein; Spgp
Rabbit
Ammonium sulfate precipitation, Size-exclusion - Dialysis
RUO
27413, 8647
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin), Western Blot
2 mg/mL
PBS with 0.09% sodium azide; pH 7.4
O95342, Q9QY30
ABCB11
KLH conjugated synthetic peptide between 1038-1067 amino acids from the C-terminal region of human ABCB11.
400 μL
Primary
Human, Mouse
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.