Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ABCB4 Monoclonal Antibody (1808CT346.21.23)
GREENER_CHOICE

Catalog No. PIMA537748
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA537748 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA537748 Supplier Invitrogen™ Supplier No. MA537748
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

ABCB4 Monoclonal Antibody for Western Blot

The membrane-associated protein encoded ABCB4 is a member of the superfamily of ATP-binding cassette (ABC) transporters. ABC proteins transport various molecules across extra- and intra-cellular membranes. ABC genes are divided into seven distinct subfamilies (ABC1, MDR/TAP, MRP, ALD, OABP, GCN20, White). This protein is a member of the MDR/TAP subfamily. Members of the MDR/TAP subfamily are involved in multidrug resistance as well as antigen presentation. This gene encodes a full transporter and member of the p-glycoprotein family of membrane proteins with phosphatidylcholine as its substrate. The function of this protein has not yet been determined; however, it may involve transport of phospholipids from liver hepatocytes into bile. Alternative splicing of this gene results in several products of undetermined function.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ABCB4
Applications Western Blot
Classification Monoclonal
Clone 1808CT346.21.23
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene ABCB4
Gene Accession No. P21439
Gene Alias ABC20; ABC21; ABCB4; ATP binding cassette subfamily B member 4; ATP-binding cassette sub-family B (MDR/TAP) member 4 (P-glycoprotein 3/ multidrug resistance 2; ATP-binding cassette sub-family B (MDR/TAP) member 4 (P-glycoprotein 3/ multidrug resistance 2); ATP-binding cassette sub-family B member 4; ATP-binding cassette, subfamily B (MDR/TAP), member 4; ATP-binding cassette, sub-family B (MDR/TAP), member 4; ATP-binding cassette, sub-family B (MDR/TAP), member 4 (P-glycoprotein 3/ multidrug resistance 2); ATP-binding cassette, subfamily B, member 4; CD243; CLCS; GBD1; GP170; ICP3; MDR1; MDR2; mdr-2; MDR2/3; MDR3; MGC163296; Multidrug resistance protein 2; multidrug resistance protein 3; multiple drug resistance 3; P glycoprotein 2; P glycoprotein 3/multiple drug resistance 3; PFIC-3; P-glycoprotein 2; P-glycoprotein 3; P-glycoprotein 3/ multidrug resistance 2; P-glycoprotein-3/multiple drug resistance-3; P-GP; Pgp3; PGY1; Pgy2; Pgy-2; Pgy3; Phosphatidylcholine translocator ABCB4
Gene Symbols ABCB4
Host Species Mouse
Immunogen recombinant protein of human ABCB4.
Purification Method Protein G
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5244
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.