Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ABCB4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA562769
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA562769 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA562769 Supplier Invitrogen™ Supplier No. PA562769
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LEAAKNGTAW RPTSAEGDFE LGISSKQKRK KTKTVKMIGV LTLFRYSDWQ D Highest antigen sequence identity to the following orthologs: Mouse - 67%, Rat - 67%.

The membrane-associated protein encoded ABCB4 is a member of the superfamily of ATP-binding cassette (ABC) transporters. ABC proteins transport various molecules across extra- and intra-cellular membranes. ABC genes are divided into seven distinct subfamilies (ABC1, MDR/TAP, MRP, ALD, OABP, GCN20, White). This protein is a member of the MDR/TAP subfamily. Members of the MDR/TAP subfamily are involved in multidrug resistance as well as antigen presentation. This gene encodes a full transporter and member of the p-glycoprotein family of membrane proteins with phosphatidylcholine as its substrate. The function of this protein has not yet been determined; however, it may involve transport of phospholipids from liver hepatocytes into bile. Alternative splicing of this gene results in several products of undetermined function.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ABCB4
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ABCB4
Gene Accession No. P21439
Gene Alias ABC20; ABC21; ABCB4; ATP binding cassette subfamily B member 4; ATP-binding cassette sub-family B (MDR/TAP) member 4 (P-glycoprotein 3/ multidrug resistance 2; ATP-binding cassette sub-family B (MDR/TAP) member 4 (P-glycoprotein 3/ multidrug resistance 2); ATP-binding cassette sub-family B member 4; ATP-binding cassette, subfamily B (MDR/TAP), member 4; ATP-binding cassette, sub-family B (MDR/TAP), member 4; ATP-binding cassette, sub-family B (MDR/TAP), member 4 (P-glycoprotein 3/ multidrug resistance 2); ATP-binding cassette, subfamily B, member 4; CD243; CLCS; GBD1; GP170; ICP3; MDR1; MDR2; mdr-2; MDR2/3; MDR3; MGC163296; Multidrug resistance protein 2; multidrug resistance protein 3; multiple drug resistance 3; P glycoprotein 2; P glycoprotein 3/multiple drug resistance 3; PFIC-3; P-glycoprotein 2; P-glycoprotein 3; P-glycoprotein 3/ multidrug resistance 2; P-glycoprotein-3/multiple drug resistance-3; P-GP; Pgp3; PGY1; Pgy2; Pgy-2; Pgy3; Phosphatidylcholine translocator ABCB4
Gene Symbols ABCB4
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ABCB4. Recombinant protein control fragment (Product #RP-101086).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5244
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.