Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ABCB5 Antibody, Alexa Fluor™ 750, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP177689AF750
Description
ABCB5 Polyclonal specifically detects ABCB5 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
ABCB5 | |
Polyclonal | |
Immunocytochemistry/Immunofluorescence | |
ABCB5 P-gp, ABCB5alpha, ABCB5beta, ATP-binding cassette protein, ATP-binding cassette sub-family B member 5, ATP-binding cassette, sub-family B (MDR/TAP), member 5, EC 3.6.3, EC 3.6.3.44, EST422562, P-glycoprotein ABCB5 | |
Rabbit | |
Affinity Purified | |
ABC Transporters, Lipid and Metabolism | |
340273.0 | |
Store at 4°C in the dark. |
Immunocytochemistry, Immunofluorescence | |
Alexa Fluor 750 | |
50 mM sodium borate with 0.05% sodium azide | |
ABCB5 | |
A synthetic peptide made to an internal portion of the human ABCB5 protein (between residues 450-500) [UniProt Q2M3G0] | |
0.1 mL | |
Primary | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction