Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ABCB5 Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody
$206.50 - $499.50
Specifications
Antigen | ABCB5 |
---|---|
Concentration | 1.11 mg/mL |
Dilution | Immunocytochemistry/Immunofluorescence 1:100 |
Applications | Immunocytochemistry, Immunofluorescence |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP177689SS
![]() |
Novus Biologicals
NBP177689SS |
0.025 mL |
Each for $206.50
|
|
|||||
NBP177689
![]() |
Novus Biologicals
NBP177689 |
0.1 mL |
Each for $499.50
|
|
|||||
Description
ABCB5 Polyclonal specifically detects ABCB5 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
ABCB5 | |
Immunocytochemistry/Immunofluorescence 1:100 | |
Polyclonal | |
Rabbit | |
ABC Transporters, Lipid and Metabolism | |
PBS and 30% Glycerol with 0.05% Sodium Azide | |
ABCB5 P-gp, ABCB5alpha, ABCB5beta, ATP-binding cassette protein, ATP-binding cassette sub-family B member 5, ATP-binding cassette, sub-family B (MDR/TAP), member 5, EC 3.6.3, EC 3.6.3.44, EST422562, P-glycoprotein ABCB5 | |
ABCB5 | |
IgG | |
Affinity Purified |
1.11 mg/mL | |
Immunocytochemistry, Immunofluorescence | |
Unconjugated | |
RUO | |
Human | |
Q2M3G0 | |
340273 | |
A synthetic peptide made to an internal portion of the human ABCB5 protein (between residues 450-500) [UniProt Q2M3G0] | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title