Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ABCB6 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA561296
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA561296 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA561296 Supplier Thermo Scientific Supplier No. PA561296
Add to Cart
Add to Cart

ABCB6 Polyclonal Antibody

ABCB6 Polyclonal Antibody for ICC/IF, IHC

TRUSTED_SUSTAINABILITY

Specifications

Antigen ABCB6
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ABCB6
Gene Accession No. Q9NP58
Gene Alias 1200005B17Rik; ABC; ABC14; ABCB6; ATG9 autophagy related 9 homolog A; ATP binding cassette subfamily B member 6; ATP binding cassette subfamily B member 6 (Langereis blood group); ATP-binding cassette half-transporter; ATP-binding cassette sub-family B member 6, mitochondrial; ATP-binding cassette, subfamily B (MDR/TAP), member 6; ATP-binding cassette, sub-family B (MDR/TAP), member 6; ATP-binding cassette, sub-family B (MDR/TAP), member 6 (Langereis blood group); ATP-binding cassette, sub-family B, member 6; DUH3; EST45597; FLJ22414; LAN; LOW QUALITY PROTEIN: ATP-binding cassette sub-family B member 6, mitochondrial; MCOPCB7; Mitochondrial ABC transporter 3; mitochondrial ATP-binding cassette sub-family B member 6; mt-ABC transporter 3; MTABC3; P-glycoprotein-related protein; PRP; PSHK2; ubiquitously-expressed mammalian ABC half transporter; Umat
Gene Symbols ABCB6
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ABCB6. Recombinant protein control fragment (Product #RP-95788).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10058
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.