Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ABCB6 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA578693
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA578693 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA578693 Supplier Invitrogen™ Supplier No. PA578693
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: HELA whole cell, MCF-7 whole cell, A549 whole cell. IHC: Human Mammary Cancer Tissue.

The membrane-associated protein encoded by this gene is a member of the superfamily of ATP-binding cassette transporters. ABC proteins transport various molecules across extra- and intra-cellular membranes. ABC genes are divided into seven distinct subfamilies. This protein is a member of the MDR/TAP subfamily. Members of the MDR/TAP subfamily are involved in multidrug resistance as well as antigen presentation. This half-transporter likely plays a role in mitochondrial function. Localized to 2q26, this gene is considered a candidate gene for lethal neonatal metabolic syndrome, a disorder of mitochondrial function.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ABCB6
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 500 μg/mL
Conjugate Unconjugated
Formulation PBS with 5mg BSA and 0.05mg sodium azide, 0.05mg thimerosal
Gene ABCB6
Gene Accession No. Q9NP58
Gene Alias 1200005B17Rik; ABC; ABC14; ABCB6; ATG9 autophagy related 9 homolog A; ATP binding cassette subfamily B member 6; ATP binding cassette subfamily B member 6 (Langereis blood group); ATP-binding cassette half-transporter; ATP-binding cassette sub-family B member 6, mitochondrial; ATP-binding cassette, subfamily B (MDR/TAP), member 6; ATP-binding cassette, sub-family B (MDR/TAP), member 6; ATP-binding cassette, sub-family B (MDR/TAP), member 6 (Langereis blood group); ATP-binding cassette, sub-family B, member 6; DUH3; EST45597; FLJ22414; LAN; LOW QUALITY PROTEIN: ATP-binding cassette sub-family B member 6, mitochondrial; MCOPCB7; Mitochondrial ABC transporter 3; mitochondrial ATP-binding cassette sub-family B member 6; mt-ABC transporter 3; MTABC3; P-glycoprotein-related protein; PRP; PSHK2; ubiquitously-expressed mammalian ABC half transporter; Umat
Gene Symbols ABCB6
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence at the C-terminus of human ABCB6 (818-842aa YADMWQLQQGQEETSEDTKPQTMER).
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10058
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.