Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ABCB6Rabbit anti-Canine, Human, Polyclonal, Rockland™
SDP

Catalog No. 501991881
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-199-1881 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-199-1881 Supplier Rockland Immunochemicals Supplier No. 600401945
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Store vial at -20°C prior to opening. Aliquot contents and freeze at -20°C or below for extended storage. Avoid cycles of freezing and thawing. Centrifuge product if not completely clear after standing at room temperature. This product is stable for several weeks at 4°C as an undiluted liquid. Dilute only prior to immediate use. This affinity-purified antibody is directed against human ABCB6 protein. A BLAST analysis was used to suggest cross reactivity with ABCB6 protein from human and dog based on 100% homology with the immunizing sequence. Expect partial reactivity with ABCB6 from mouse and rat sources based on partial (∼93%, 15/16) sequence homology. Reactivity against homologues from other sources is not known.

The membrane-associated protein encoded by this gene is a member of the superfamily of ATP-binding cassette transporters. ABC proteins transport various molecules across extra- and intra-cellular membranes. ABC genes are divided into seven distinct subfamilies. This protein is a member of the MDR/TAP subfamily. Members of the MDR/TAP subfamily are involved in multidrug resistance as well as antigen presentation. This half-transporter likely plays a role in mitochondrial function. Localized to 2q26, this gene is considered a candidate gene for lethal neonatal metabolic syndrome, a disorder of mitochondrial function.

Specifications

Antigen ABCB6
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 0.02M potassium phosphate with 0.15M NaCl and 0.01% sodium azide; pH 7.2
Gene ABCB6
Gene Accession No. Q9NP58
Gene Alias 1200005B17Rik; ABC; ABC14; Abcb6; ATG9 autophagy related 9 homolog A; ATG9A; ATP binding cassette subfamily B member 6 (Langereis blood group); ATP-binding cassette half-transporter; ATP-binding cassette sub-family B member 6, mitochondrial; ATP-binding cassette, subfamily B (MDR/TAP), member 6; ATP-binding cassette, sub-family B (MDR/TAP), member 6; ATP-binding cassette, sub-family B (MDR/TAP), member 6 (Langereis blood group); ATP-binding cassette, sub-family B, member 6; autophagy related 9A; BOS_2119; DUH3; EST45597; FLJ22414; LAN; MCOPCB7; Mitochondrial ABC transporter 3; mitochondrial ATP-binding cassette sub-family B member 6; Mt-ABC transporter 3; MTABC3; P-glycoprotein-related protein; PRP; PSHK2; Ubiquitously-expressed mammalian ABC half transporter; UMAT
Gene Symbols ABCB6
Host Species Rabbit
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10058, 478914
Target Species Canine, Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.