Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ABCB9 Monoclonal Antibody (4F4)
GREENER_CHOICE

Catalog No. PIMA521478
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA521478 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA521478 Supplier Invitrogen™ Supplier No. MA521478
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Peptide Sequence: FIDRQPTMVH DGSLAPDHLE GRVDFENVTF TYRTRPHTQV LQNVSFSLSP GKVTALVGPS GSGKSSCVNI LENFYPLEGG RVLLDGKPIS AYDHKYLHR.

The structure specific ERCC 1 / XPF endonuclease complex is implicated in the repair of two distinct types of lesions in DNA: NER for UV induced lesions and bulky chemical adducts; and recombination repair of the very genotoxic interstrand cross links. NER mechanism involves dual incisions on both sides of the damage catalyzed by two nucleases. In mammalian cells, XPG cleaves 3' of the DNA lesion while the ERCC1 / XPF complex makes the 5' incision. ERCC1 / XPF probably plays a role in processing partially processed recombination intermediates.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ABCB9
Applications ELISA, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 4F4
Concentration 1.0 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene ABCB9
Gene Accession No. Q9NP78
Gene Alias ABC transporter 9 protein; ABCB9; ATP binding cassette subfamily B member 9; ATP-binding cassette sub-family B member 9; ATP-binding cassette transporter 9; ATP-binding cassette, subfamily B (MDR/TAP), member 9; ATP-binding cassette, sub-family B (MDR/TAP), member 9; ATP-binding cassette, sub-family B, member 9; EST122234; KIAA1520; LOW QUALITY PROTEIN: ATP-binding cassette sub-family B member 9; mABCB9; mKIAA1520; TAPL; TAP-like ABC transporter; TAP-like protein
Gene Symbols ABCB9
Host Species Mouse
Immunogen ABCB9 (NP_062571, 482 a.a. ~ 580 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23457
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.