Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ABCB9 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501981775
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-1775 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-1775 Supplier Bioss Supplier No. BS11907R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The structure specific ERCC 1 / XPF endonuclease complex is implicated in the repair of two distinct types of lesions in DNA: NER for UV induced lesions and bulky chemical adducts; and recombination repair of the very genotoxic interstrand cross links. NER mechanism involves dual incisions on both sides of the damage catalyzed by two nucleases. In mammalian cells, XPG cleaves 3′ of the DNA lesion while the ERCC1 / XPF complex makes the 5′ incision. ERCC1 / XPF probably plays a role in processing partially processed recombination intermediates.

Specifications

Antigen ABCB9
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene ABCB9
Gene Accession No. Q9NP78
Gene Alias ABC transporter 9 protein; ABCB9; ATP binding cassette subfamily B member 9; ATP-binding cassette sub-family B member 9; ATP-binding cassette transporter 9; ATP-binding cassette, subfamily B (MDR/TAP), member 9; ATP-binding cassette, sub-family B (MDR/TAP), member 9; ATP-binding cassette, sub-family B, member 9; BOS_16586; EST122234; KIAA1520; mABCB9; mKIAA1520; TAPL; TAP-like ABC transporter; TAP-like protein
Gene Symbols ABCB9
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human ABCB9.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23457
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.