Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ABCC11 Monoclonal Antibody (MRP8 7C12/4), Invitrogen™

Catalog No. PIMA190712
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA190712 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA190712 Supplier Thermo Scientific Supplier No. MA190712

Mouse Monoclonal Antibody

Western blot detects a band ∽82 kDa. A suggested positive control for this product is neutrophil lysate (particularly neutrophils from spleen).

ABCC11 Monoclonal antibody specifically detects ABCC11 in Human samples. It is validated for ELISA, Flow Cytometry, Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ABCC11
Applications ELISA, Flow Cytometry, Western Blot
Classification Monoclonal
Clone MRP8 7C12/4
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene ABCC11
Gene Accession No. Q96J66
Gene Alias ABCC11; ATP binding cassette subfamily C member 11; ATP-binding cassette protein C11; ATP-binding cassette sub-family C member 11; ATP-binding cassette transporter MRP8; ATP-binding cassette transporter sub-family C member 11; ATP-binding cassette, sub-family C (CFTR/MRP), member 11; EWWD; MRP8; multidrug resistance-associated protein 8; multi-resistance protein 8; WW
Gene Symbols ABCC11
Host Species Mouse
Immunogen recombinant Human MRP8
Purification Method IgM
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 85320
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgM
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.