Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ABCC4 Mouse anti-Human, Clone:6A7H3, Abnova™
Mouse monoclonal antibody raised against recombinant human ABCC4.
Supplier: Abnova Corporation MAB17767
Description
The protein encoded by this gene is a member of the superfamily of ATP-binding cassette (ABC) transporters. ABC proteins transport various molecules across extra- and intra-cellular membranes. ABC genes are divided into seven distinct subfamilies (ABC1, MDR/TAP, MRP, ALD, OABP, GCN20, White). This protein is a member of the MRP subfamily which is involved in multi-drug resistance. The specific function of this protein has not yet been determined; however, this protein may play a role in cellular detoxification as a pump for its substrate, organic anions. Alternative splicing results in multiple splice variants encoding different isoforms. [provided by RefSeq]Specifications
ABCC4 | |
Monoclonal | |
Unconjugated | |
ELISA (1:10000) Flow Cytometry (1:200-1:400) Immunocytochemistry Immunohistochemistry (1:200-1:1000) Western Blot (1:500-1:2000) The optimal working dilution should be determined by the end user. |
|
ABCC4 | |
Mouse | |
100 μg | |
Human | |
Antibody | |
IgG1 |
ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
6A7H3 | |
ATP-binding cassette, sub-family C (CFTR/MRP), member 4 | |
In PBS (0.05% sodium azide) | |
EST170205, MOAT-B, MOATB, MRP4 | |
Recombinant protein corresponding to amino acids 631-692 of human ABCC4 from E. coli. | |
10257 | |
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction