Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ABCC5 Antibody (OTI2C8), FITC, Novus Biologicals™
SDP

Catalog No. NBP271540F Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP271540F 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP271540F Supplier Novus Biologicals Supplier No. NBP271540F
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

ABCC5 Monoclonal specifically detects ABCC5 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry.

Specifications

Antigen ABCC5
Applications Immunohistochemistry
Classification Monoclonal
Clone OTI2C8
Conjugate FITC
Dilution Immunohistochemistry
Formulation PBS with 0.05% Sodium Azide
Gene Alias ATP-binding cassette sub-family C member 5, ATP-binding cassette, sub-family C (CFTR/MRP), member 5, canalicular multispecific organic anion transporter C, EST277145, MOAT-CABC33, MRP5MOATC, multidrug resistance-associated protein 5, Multi-specific organic anion transporter C, pABC11, SMRPDKFZp686C1782
Gene Symbols ABCC5
Host Species Mouse
Immunogen Full length recombinant protein of human ABCC5 (NP_001018881) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline ABC Transporters, Amino Acids Drugs and other small molecules, Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 10057.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.