Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ABCC5 Antibody (OTI2C8), PE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP271540PE
Description
ABCC5 Monoclonal antibody specifically detects ABCC5 in Human, Mouse, Rat samples. It is validated for Western Blot, ImmunohistochemistrySpecifications
ABCC5 | |
Monoclonal | |
PE | |
PBS | |
Mouse | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Western Blot, Immunohistochemistry | |
OTI2C8 | |
Western Blot, Immunohistochemistry | |
ATP-binding cassette sub-family C member 5, ATP-binding cassette, sub-family C (CFTR/MRP), member 5, canalicular multispecific organic anion transporter C, EST277145, MOAT-CABC33, MRP5MOATC, multidrug resistance-associated protein 5, Multi-specific organic anion transporter C, pABC11, SMRPDKFZp686C1782 | |
Full length human recombinant protein of human ABCC5(NP_001018881) produced in E.coli. | |
0.1 mL | |
ABC Transporters, Amino Acids Drugs and other small molecules, Lipid and Metabolism | |
10057 | |
Store at 4C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction