Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Abcc8 Mouse anti-Human, Rat, FITC, Clone:S289-16, Abnova™
Mouse monoclonal antibody raised against partial recombinant rat Abcc8.
Supplier: Abnova Corporation MAB16806
Specifications
Abcc8 | |
Monoclonal | |
FITC | |
Immunocytochemistry (1:100) Immunofluorescence (1:100) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:1000) Western Blot (1:1000) The optimal working dilution should be determined by the end user. |
|
Abcc8 | |
Mouse | |
100 μg | |
Human, Rat | |
Antibody | |
IgG1 |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
S289-16 | |
ATP-binding cassette, sub-family C (CFTR/MRP), member 8 | |
In PBS, pH 7.4 (50% glycerol, 0.09% sodium azide) | |
Sur, Sur1 | |
Recombinant protein corresponding to amino acids 1548-1582 at C-terminus of rat Abcc8. | |
25559 | |
Store at -20°C. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction