Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Abcc9 Mouse anti-Human, Mouse, Rat, Biotin, Clone:S319A-14, Abnova™
Mouse monoclonal antibody raised against partial recombinant mouse Abcc9.
Supplier: Abnova Corporation MAB16841
Specifications
Abcc9 | |
Monoclonal | |
Biotin | |
Immunocytochemistry Immunofluorescence Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) Western Blot (1:1000) The optimal working dilution should be determined by the end user. |
|
Abcc9 | |
Mouse | |
100 μg | |
Human, Mouse, Rat | |
Antibody | |
IgG2a |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
S319A-14 | |
ATP-binding cassette, sub-family C (CFTR/MRP), member 9 | |
In PBS, pH 7.4 (50% glycerol, 0.09% sodium azide) | |
AI414027, AI449286, SUR2A, SUR2B, Sur2 | |
Recombinant protein corresponding to amino acids 1505-1546 at C-terminus of mouse Abcc9. | |
20928 | |
Store at -20°C. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction