Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ABCC9 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5103640
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5103640 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5103640 Supplier Invitrogen™ Supplier No. PA5103640
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total ABCC9.

Sulfonylurea receptors (SUR) are membrane proteins which are the molecular targets of the sulfonylurea class of anti-diabetic drugs whose mechanism of action is to promote insulin release from pancreatic beta cells. More specifically, SUR proteins are subunits of the inward-rectifier potassium ion channels Kir6.x (6.1 and 6.2). The association of four Kir6.x and four SUR subunits form an ion conducting channel commonly referred to as the KATP channel. The primary function of the sulfonylurea receptor is to sense intracellular levels of the nucleotides ATP and ADP and in response facilitate the open or closing its associated Kir6.x potassium channel. Hence the KATP channel monitors the energy balance within the cell.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ABCC9
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene ABCC9
Gene Accession No. O60706, Q63563
Gene Alias ABC37; ABCC9; AI414027; AI449286; ATFB12; ATP binding cassette subfamily C member 9; ATP-binding cassette sub-family C member 9; ATP-binding cassette transporter sub-family C member 9; ATP-binding cassette, subfamily C (CFTR/MRP), member 9; ATP-binding cassette, sub-family C (CFTR/MRP), member 9; CANTU; cardiac ventricle sulfonyl urea receptor; CMD1O; FLJ36852; membrane receptor protein; si:dkey-183c2.3; Sulfonylurea receptor 2; sulfonylurea receptor subunit 2; sulfonylurea-binding protein 2; sulphonylurea receptor 2; sulphonylurea receptor 2A; sulphonylurea receptor 2B; SUR2; SUR2A; SUR2B
Gene Symbols ABCC9
Host Species Rabbit
Immunogen A synthesized peptide derived from human ABCC9(Accession O60706), corresponding to amino acid residues L1269-V1319.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10060, 25560
Target Species Human, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.