Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ABCD1 Antibody (1AL-2B4), Novus Biologicals™
SDP

Catalog No. p-7201155 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB019438 0.025 mL
NBP229709 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB019438 Supplier Novus Biologicals Supplier No. NBP2297090.025ML
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

ABCD1 Monoclonal specifically detects ABCD1 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen ABCD1
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry, ELISA, Western Blot
Classification Monoclonal
Clone 1AL-2B4
Conjugate Unconjugated
Dilution Western Blot 1:500-1:5000, ELISA 1:500-1:5000, Immunohistochemistry 1:500-1:5000, Immunocytochemistry/Immunofluorescence 1:500-1:5000
Formulation Ascites with No Preservative
Gene Accession No. P33897
Gene Alias Adrenoleukodystrophy protein, ALDATP-binding cassette sub-family D member 1, ALDPadrenoleukodystrophy, AMNABC42, ATP-binding cassette, sub-family D (ALD), member 1
Gene Symbols ABCD1
Host Species Mouse
Immunogen ALDP fragment from aa 495-648 as a fusion protein.
Purification Method Unpurified
Quantity 0.025 mL
Regulatory Status RUO
Research Discipline ABC Transporters, Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 215
Test Specificity Human adrenoleukodystrophy protein (ALDP). No cross-reactivity on mouse ALDP, mouse ALDRP and PMP 70. MAB2162 can be used in Western Blot, immunofluorescence or immunohistochemical studies to analyze the ALD protein in cells and tissues from normal individuals or from patients with adrenoleukodystrophy (ALD) or Zellweger syndrome and related peroxisomal diseases, and provides an excellent marker for peroxisomes(Mosser et al., 1994). About 70% of ALD male patients have no detectable ALDP in fibroblasts or leukocytes. A similar proportion of carrier females may show mosaicism for the presence or absence of ALDP.
Target Species Human
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Form Ascites
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.