Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ABCD1 Mouse anti-Human, Clone: 1H8E5, Proteintech

Mouse Monoclonal Antibody
$518.60
Specifications
Antigen | ABCD1 |
---|---|
Clone | 1H8E5 |
Concentration | 1 mg/mL |
Applications | Immunocytochemistry, Immunofluorescence, Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-6267
![]() ![]() |
Proteintech Group Inc
601531IG150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
This gene is a member of the UBA domain family, whose members include proteins having connections to ubiquitin and the ubiquitination pathway. The ubiquitin associated domain is thought to be a non-covalent ubiquitin binding domain consisting of a compact three helix bundle. This particular protein originates from a gene locus in a refined region on chromosome 9 undergoing loss of heterozygosity in nasopharyngeal carcinoma (NPC). Taking into account its cytogenetic location, this UBA domain family member is being studies as a putative target for mutation in nasopharyngeal carcinomas. Multiple alternatively spliced transcript variants encoding different isoforms have been found for this gene.Specifications
ABCD1 | |
1 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
ABC42, ABCD1, Adrenoleukodystrophy protein, ALD, ALDP, AMN | |
ABCD1 | |
IgG1 | |
Caprylic Acid Precipitation | |
Antibody |
1H8E5 | |
Immunocytochemistry, Immunofluorescence, Western Blot | |
Unconjugated | |
Mouse | |
Human | |
P33897 | |
215 | |
ABCD1 Fusion Protein Ag12757 | |
Primary | |
-20°C | |
ABCD1 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title