Learn More
Invitrogen™ ABCD2 Polyclonal Antibody
Description
ABCD2 Polyclonal Antibody for Western Blot, IHC (P)
Specifications
Specifications
| Antigen | ABCD2 |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 |
| Gene | ABCD2 |
| Gene Accession No. | Q61285, Q9UBJ2 |
| Gene Alias | ABC39; Abcd2; adrenoleukodystrophy related; adrenoleukodystrophy-like 1; Adrenoleukodystrophy-related protein; ALD1; ALDL1; ALDR; ALDRP; ATP binding cassette subfamily D member 2; ATP-binding cassette sub-family D member 2; ATP-binding cassette, subfamily D (ALD), member 2; ATP-binding cassette, sub-family D (ALD), member 2; ATP-binding cassette, sub-family D, member 2; hALDR; LOW QUALITY PROTEIN: ATP-binding cassette sub-family D member 2 |
| Gene Symbols | ABCD2 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.