Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ABCD2 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501980951
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-0951 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-0951 Supplier Bioss Supplier No. BS10646R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ABCD2 is a member of the superfamily of ATP-binding cassette (ABC) transporters. ABC proteins transport various molecules across extra- and intra-cellular membranes. ABC genes are divided into seven distinct subfamilies (ABC1, MDR/TAP, MRP, ALD, OABP, GCN20, White). This protein is a member of the ALD subfamily, which is involved in peroxisomal import of fatty acids and/or fatty acyl-CoAs in the organelle. All known peroxisomal ABC transporters are half transporters which require a partner half transporter molecule to form a functional homodimeric or heterodimeric transporter. The function of this peroxisomal membrane protein is unknown; however this protein is speculated to function as a dimerization partner of ABCD1 and/or other peroxisomal ABC transporters.

Specifications

Antigen ABCD2
Applications Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene ABCD2
Gene Accession No. Q9UBJ2
Gene Alias ABC39; Abcd2; adrenoleukodystrophy related; Adrenoleukodystrophy-like 1; Adrenoleukodystrophy-related protein; ALD1; ALDL1; Aldr; Aldrp; ATP binding cassette subfamily D member 2; ATP-binding cassette sub-family D member 2; ATP-binding cassette, subfamily D (ALD), member 2; ATP-binding cassette, sub-family D (ALD), member 2; ATP-binding cassette, sub-family D, member 2; BOS_5215; hALDR; LOW QUALITY PROTEIN: ATP-binding cassette sub-family D member 2
Gene Symbols ABCD2
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human ABCD2.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 225
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.