Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ABCD4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5109352
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5109352 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5109352 Supplier Invitrogen™ Supplier No. PA5109352
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: DQFTCNLLYV SWRKDLTEHL HRLYFRGRAY YTLNVLRDDI DNPDQRISQD VERFCRQLSS MASKLIISPF TLVYYTYQCF QSTGWLGPVS IFGYFILGTV VNKTLMGPIV MKLVHQEKLE GDFRFKHMQI RVNAEPAAFY RAGHVEHMRT DRRLQRLLQT QRELMSKELW LYIGINTFDY L.

The peroxisomal membrane contains several ATP-binding cassette (ABC) transporters, ABCD1-4 that are known to be present in the human peroxisome membrane. All four proteins are ABC half-transporters, which dimerize to form an active transporter. A mutation in the ABCD1 gene causes X-linked adreno-leukodystrophy (X-ALD), a peroxisomal disorder which affects lipid storage. ABCD2 in mouse is expressed at high levels in the brain and adrenal organs, which are adversely affected in X-ALD. The peroxisomal membrane comprises two quantitatively major proteins, PMP22 and ABCD3. ABCD3 is associated with irregularly shaped vesicles which may be defective peroxisomes or peroxisome precursors. ABCD1 localizes to peroxisomes. ABCB7 is a half-transporter involved in the transport of heme from the mitochondria to the cytosol.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ABCD4
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.41 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene ABCD4
Gene Accession No. O14678
Gene Alias 69 kDa peroxisomal ABC-transporter; ABC41; ABCD4; ATP binding cassette subfamily D member 4; ATP-binding cassette sub-family D member 4; ATP-binding cassette, subfamily D (ALD), member 4; ATP-binding cassette, sub-family D (ALD), member 4; ATP-binding cassette, sub-family D, member 4; EST352188; MAHCJ; P69r; P70R; P79R; Peroxisomal membrane protein 1-like; Peroxisomal membrane protein 69; peroxisomal membrane protein, 70 kDa-related; PMP69; PMP70-related protein; Pxmp1l; PXMP1-L
Gene Symbols ABCD4
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 100-280 of human ABCD4 (NP_005041.1).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5826
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.