Learn More
Invitrogen™ ABCE1 Polyclonal Antibody

Description
The antibody detects endogenous levels of total ABCE1 protein.
Specifications
Specifications
| Antigen | ABCE1 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 1.1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 |
| Gene | ABCE1 |
| Gene Accession No. | P61221, P61222 |
| Gene Alias | 2'-5'-oligoadenylate-binding protein; ABC 38; ABC38; ABCE 1; ABCE1; ATP binding cassette subfamily E member 1; ATP-binding cassette sub-family E member 1; ATP-binding cassette, subfamily E (OABP), member 1; ATP-binding cassette, sub-family E (OABP), member 1; ATP-binding cassette, subfamily E, member 1; C79080; HP68; HuHP68; OABP; OK/SW-cl.40; Ribonuclease 4 inhibitor; ribonuclease L (2', 5'-oligoisoadenylate synthetase-dependent) inhibitor; ribonuclease L (2',5'-oligoisoadenylate synthetase-dependent) inhibitor; RLI; RLI1; RNase 4 inhibitor; RNase L inhibitor; RNase L inhibitor 1; RNASEL1; Rnaseli; RNS41; RNS4I; RNS4l (Eye) |
| Gene Symbols | ABCE1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.