Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ABCE1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP30495920UL
Description
ABCE1 Polyclonal specifically detects ABCE1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
ABCE1 | |
Polyclonal | |
PBS with 50% glycerol, pH7.3. | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Western Blot, Immunofluorescence | |
Unconjugated | |
2'-5'-oligoadenylate-binding protein, ATP-binding cassette, sub-family E (OABP), member 1, HuHP68, OABP, Ribonuclease 4 inhibitor, ribonuclease L (2'-5'-oligoisoadenylate synthetase-dependent) inhibitor, RLIABC38, RNase L inhibitor, RNASEL1, RNASELIRNS4IATP-binding cassette sub-family E member 1 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 320-599 of human ABCE1 (NP_002931.2). | |
20 μL | |
ABC Transporters, Immune System Diseases, Immunology, Lipid and Metabolism, Membrane Trafficking and Chaperones, Plasma Membrane Markers, Signal Transduction | |
6059 | |
Store at -20°C. Avoid freeze-thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction