Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ABCF1 Polyclonal Antibody

Catalog No. PIPA5122296
Change view
Click to view available options
Quantity:
20 μL
200 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5122296 20 μL
PIPA5118972 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. PIPA5122296 Supplier Invitrogen™ Supplier No. PA5122296
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

ABCF1 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)

This gene encodes a component of the spliceosome complex and is one of several retinitis pigmentosa-causing genes. When the gene product is added to the spliceosome complex, activation occurs.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ABCF1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
Gene Abcf1
Gene Accession No. Q8NE71
Gene Alias ABC27; ABC50; ABCF1; ATP binding cassette subfamily F member 1; ATP-binding cassette 50; ATP-binding cassette 50 (TNF-alpha stimulated); ATP-binding cassette sub-family F member 1; ATP-binding cassette sub-family F member 1; LOW QUALITY PROTEIN: ATP-binding cassette sub-family F member 1; ATP-binding cassette, subfamily F (GCN20), member 1; ATP-binding cassette, sub-family F (GCN20), member 1; ATP-binding cassette, sub-family F, member 1; AU041969; D17Wsu166e; GCN20; TNF-alpha stimulated ABC protein; TNFalpha-inducible ATP-binding protein; TNF-alpha-stimulated ABC protein; wu:fb79c06; wu:fc39a05; wu:fj94a08; zgc:85667
Gene Symbols Abcf1
Host Species Rabbit
Immunogen Recombinant protein ATP Binding Cassette Transporter F1. The antigen corresponds to amino acid range 625-845 of the target protein.
Purification Method Antigen affinity chromatography, Protein A
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.