Learn More
Invitrogen™ ABCF1 Polyclonal Antibody

Description
Immunogen sequence: KKDVDDDGEE KELMERLKKL SVPTSDEEDE VPAPKPRGGK KTKGGNVFAA LIQDQSEEEE EEEKHPPKPA KPEKNRINKA VSEEQQPALK GKKGKEEKSK GKAKPQNKFA AL Highest antigen sequence indentity to the following orthologs: Rat - 72%, Mouse - 73%.
Specifications
Specifications
| Antigen | ABCF1 |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | Abcf1 |
| Gene Accession No. | Q8NE71 |
| Gene Alias | ABC27; ABC50; ABCF1; ATP binding cassette subfamily F member 1; ATP-binding cassette 50; ATP-binding cassette 50 (TNF-alpha stimulated); ATP-binding cassette sub-family F member 1; ATP-binding cassette sub-family F member 1; LOW QUALITY PROTEIN: ATP-binding cassette sub-family F member 1; ATP-binding cassette, subfamily F (GCN20), member 1; ATP-binding cassette, sub-family F (GCN20), member 1; ATP-binding cassette, sub-family F, member 1; AU041969; D17Wsu166e; GCN20; TNF-alpha stimulated ABC protein; TNFalpha-inducible ATP-binding protein; TNF-alpha-stimulated ABC protein; wu:fb79c06; wu:fc39a05; wu:fj94a08; zgc:85667 |
| Gene Symbols | Abcf1 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.