Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ABCF1 Rabbit anti-Human, Rat, Clone: 4P9V8, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP31581920UL
This item is not returnable.
View return policy
Description
ABCF1 Monoclonal antibody specifically detects ABCF1 in Human, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
| ABCF1 | |
| Monoclonal | |
| Unconjugated | |
| PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Rat | |
| Purified |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 4P9V8 | |
| Western Blot, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin | |
| ABC27, ABC50TNFalpha-inducible ATP-binding protein, ATP-binding cassette 50, ATP-binding cassette 50 (TNF-alpha stimulated), ATP-binding cassette sub-family F member 1, ATP-binding cassette, sub-family F (GCN20), member 1, EC 2.7.7.6, EC 3.6.3.17, EST123147, TNF-alpha-stimulated ABC protein | |
| A synthetic peptide corresponding to a sequence within amino acids 746-845 of human ABCF1 (Q8NE71). | |
| 20 μg | |
| Immunology | |
| 23 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction