Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ABCF2 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$206.50 - $499.50
Specifications
Antigen | ABCF2 |
---|---|
Dilution | Western Blot 1:500-1:1000, Simple Western 1:1000, Immunocytochemistry/Immunofluorescence 1:500 |
Applications | Western Blot, Immunocytochemistry, Immunofluorescence |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB400115SS
![]() |
Novus Biologicals
NB400115SS |
0.025 mL |
Each for $206.50
|
|
|||||
NB400115
![]() |
Novus Biologicals
NB400115 |
0.1 mL |
Each for $499.50
|
|
|||||
Description
ABCF2 Polyclonal specifically detects ABCF2 in Human, Mouse, Yeast samples. It is validated for Western Blot, Simple Western, Immunocytochemistry/Immunofluorescence, Knockdown Validated.Specifications
ABCF2 | |
Western Blot, Immunocytochemistry, Immunofluorescence | |
Unconjugated | |
Rabbit | |
ABC Transporters, Chromatin Research, DNA replication Transcription Translation and Splicing, Lipid and Metabolism, Membrane Trafficking and Chaperones, Signal Transduction | |
Whole antisera with 0.05% Sodium Azide | |
ABC28, ABC-type transport protein, ATP-binding cassette sub-family F member 2, ATP-binding cassette, sub-family F (GCN20), member 2, DKFZp586K1823, EC 3.6.3, EC 3.6.3.17, EST133090, HUSSY18, HUSSY-18, Iron inhibited ABC transporter 2, Iron-inhibited ABC transporter 2, M-ABC1 | |
ABCF2 | |
IgG | |
Unpurified | |
Human, Mouse, Yeast ABCF2. |
Western Blot 1:500-1:1000, Simple Western 1:1000, Immunocytochemistry/Immunofluorescence 1:500 | |
Polyclonal | |
Antisera | |
RUO | |
Human, Mouse, Yeast | |
Q9UG63 | |
10061 | |
A fusion protein containing amino acids 1-102 of the ABCF2 protein. | |
Primary | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. | |
71 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title