Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ABCF2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. APSA6796
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
APSA6796 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. APSA6796 Supplier Invitrogen™ Supplier No. PA316796
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Reactive to yeast homolog ARB1.

OCIAD1 was identified via immunoscreening of an ovarian carcinoma cDNA library from ovarian cancer patients and is expressed in multiple tissues including ovary, placenta, brain, testis, prostate, and mammary gland. Two isoforms of OCIAD1 are known to exist; the shorter isoform is restricted to the central nervous system. OCIAD1 is a transmembrane protein whose overexpression in HEY ovarian cancer cells increased lysophosphatidic acid- (LPA-)induced, but not basal level cell adhesion to extracellular matrix proteins collagen I and laminin10/11. This adhesion is not blocked by LY294002 and GF109203X, suggesting that OCIAD1 does not use protein kinase C and PI3 kinase signaling pathways to exert its effect on adhesion.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ABCF2
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration Conc. Not Determined
Conjugate Unconjugated
Formulation Whole serum with 0.05% sodium azide
Gene Abcf2
Gene Accession No. Q99LE6, Q9UG63
Gene Alias 0710005O05Rik; ABC28; Abcf2; ABC-type transport protein; ATP binding cassette subfamily F member 2; ATP-binding cassette sub-family F member 2; ATP-binding cassette, subfamily F (GCN20), member 2; ATP-binding cassette, sub-family F (GCN20), member 2; developmentally regulated repeat element-containing transcript 3; DKFZp586K1823; Drr3; E430001O06; EST133090; HUSSY18; HUSSY-18; Iron-inhibited ABC transporter 2; M-ABC1
Gene Symbols Abcf2
Host Species Rabbit
Immunogen A fusion protein containing amino acids 1-102 of the ABCF2 protein.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10061, 27407
Target Species Human, Mouse, Yeast
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.