Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ABCF2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA556456
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA556456 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA556456 Supplier Invitrogen™ Supplier No. PA556456
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: CVWLEEELKT FKRILVLVSH SQDFLNGVCT NIIHMHNKKL KYYTGNYDQY VKTRLELEEN QMKRFHWEQD Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.

OCIAD1 was identified via immunoscreening of an ovarian carcinoma cDNA library from ovarian cancer patients and is expressed in multiple tissues including ovary, placenta, brain, testis, prostate, and mammary gland. Two isoforms of OCIAD1 are known to exist; the shorter isoform is restricted to the central nervous system. OCIAD1 is a transmembrane protein whose overexpression in HEY ovarian cancer cells increased lysophosphatidic acid- (LPA-)induced, but not basal level cell adhesion to extracellular matrix proteins collagen I and laminin10/11. This adhesion is not blocked by LY294002 and GF109203X, suggesting that OCIAD1 does not use protein kinase C and PI3 kinase signaling pathways to exert its effect on adhesion.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ABCF2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Abcf2
Gene Accession No. Q99LE6, Q9UG63
Gene Alias 0710005O05Rik; ABC28; Abcf2; ABC-type transport protein; ATP binding cassette subfamily F member 2; ATP-binding cassette sub-family F member 2; ATP-binding cassette, subfamily F (GCN20), member 2; ATP-binding cassette, sub-family F (GCN20), member 2; developmentally regulated repeat element-containing transcript 3; DKFZp586K1823; Drr3; E430001O06; EST133090; HUSSY18; HUSSY-18; Iron-inhibited ABC transporter 2; M-ABC1
Gene Symbols Abcf2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ABCF2. Recombinant protein control fragment (Product #RP-92615).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10061, 27407, 311959
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.