Learn More
Invitrogen™ ABCF2 Polyclonal Antibody

Description
Positive test controls include: 22Rv1, 293T, HeLa, MCF7, Mouse testis, Mouse skeletal muscle, Mouse brain, Rat intestine. Immunogen sequence: MPSDLAKKKA AKKKEAAKAR QRPRKGHEEN GDVVTEPQVA EKNEANGRET TEVDLLTKEL EDFEMKKAAA RAVTGVLASH PNSTDVHIIN LSLTFHGQEL LSDTKLELNS GRRYGLIGLN GIGKSMLLSA IGKREVPIPE HIDIYHLTRE MPPSDKTPLH CVMEVDTERA MLEKEAERLA HEDAECEKLM ELYERLEELD ADKAEMRASR ILHGLGFTPA MQRKKLKDFS GGWRMRVALA RALFIRPFML.
Specifications
Specifications
| Antigen | ABCF2 |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Concentration | 0.99 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | Abcf2 |
| Gene Accession No. | Q99LE6, Q9UG63 |
| Gene Alias | 0710005O05Rik; ABC28; Abcf2; ABC-type transport protein; ATP binding cassette subfamily F member 2; ATP-binding cassette sub-family F member 2; ATP-binding cassette, subfamily F (GCN20), member 2; ATP-binding cassette, sub-family F (GCN20), member 2; developmentally regulated repeat element-containing transcript 3; DKFZp586K1823; Drr3; E430001O06; EST133090; HUSSY18; HUSSY-18; Iron-inhibited ABC transporter 2; M-ABC1 |
| Gene Symbols | Abcf2 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.