Learn More
ABCF2 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
$151.31 - $568.26
Specifications
| Antigen | ABCF2 |
|---|---|
| Applications | Immunohistochemistry |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Form | Liquid |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-157-2076
|
Bethyl Laboratories, Inc
IHC00425T |
10 μL |
Each for $151.31
|
|
|||||
50-157-2075
|
Bethyl Laboratories, Inc
IHC00425 |
100 μL |
Each for $568.26
|
|
|||||
Description
The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Mouse, Rat, Rabbit, Guinea pig_10141, Orangutan, Monkey, Gorilla, Chimpanzee, Gray short-t
ABCF2 is a member of the ABC Transporter superfamily and of the GCN20 subfamily. As a member of the ABC Transporter family, it functions to transport various molecules across extra- and intracellular membranes.Specifications
| ABCF2 | |
| Polyclonal | |
| Liquid | |
| RUO | |
| TBS with 0.1% BSA and 0.09% sodium azide | |
| ABC28, DKFZp586K1823, EST133090, HUSSY-18, HUSSY18, M-ABC1 | |
| ABCF2 | |
| IgG | |
| Antigen affinity chromatography | |
| Antibody |
| Immunohistochemistry | |
| Unconjugated | |
| Rabbit | |
| Human | |
| Q9UG63 | |
| 10061 | |
| Between 573 and 623 | |
| Primary | |
| 4° C | |
| ABCF2 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.