Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ABCF3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5120333
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5120333 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5120333 Supplier Invitrogen™ Supplier No. PA5120333
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive test controls include: Jurkat, Mouse brain, Mouse lung, Rat testis. Immunogen sequence: MATCAEILRS EFPEIDGQVF DYVTGVLHSG SADFESVDDL VEAVGELLQE VSGDSKDDAG IRAVCQRMYN TLRLAEPQSQ GNSQVLLDAP IQLSKITENY DCGTKLPGLL KREQSSTVNA KKLEKAEARL KAKQEKRSEK DTLKTSNPLV LEEASASQAG SRKESRLESS GKNKSYDVRI ENFDVSFGDR VLLAGADVNL.

This protein may display an antiviral effect against flaviviruses such as west Nile virus (WNV) in the presence of OAS1B. ATP-binding cassette (ABC) transporters regulate the transport of a variety of physiologic substrates. But this protein lacks transmembrane domains and is probably not involved in transport.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ABCF3
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.72 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene ABCF3
Gene Accession No. Q66H39, Q8K268, Q9NUQ8
Gene Alias ABCF3; AI326318; ATP binding cassette subfamily F member 3; ATP-binding cassette sub-family F member 3; ATP-binding cassette, subfamily F (GCN20), member 3; ATP-binding cassette, sub-family F (GCN20), member 3; AU016058; BB119416; EST201864
Gene Symbols ABCF3
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-200 of human ABCF3 (NP_060828.2).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27406, 287982, 55324
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.